Tisk…

Anti-MAPRE2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-7837R-A647EA 24190 CZK
BOSSBS-7837R-A647
Anti-MAPRE2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. This protein is a microtubule-associated protein that is necessary for spindle symmetry during mitosis. It is thought to play a role in the tumorigenesis of colorectal cancers and the proliferative control of normal cells. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq, Jan 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAPRE2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR