Tisk…

Anti-MAD2L2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BOSSBS-7840R-A350EA 24190 CZK
BOSSBS-7840R-A350
Anti-MAD2L2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
MAD2L2 is a component of the mitotic spindle assembly checkpoint that prevents the onset of anaphase until all chromosomes are properly aligned at the metaphase plate. MAD2L2 is a homolog of MAD2L1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAD2L2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR