Tisk…

Anti-NUP160 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-7843R-CY7EA 24190 CZK
BOSSBS-7843R-CY7
Anti-NUP160 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
NUP160 is 1 of up to 60 proteins that make up the 120 MD nuclear pore complex, which mediates nucleoplasmic transport. NUP160 forms part of the Nup160 subcomplex in the nuclear pore which is composed of NUP160, Nup133, Nup107 and Nup96. This complex plays a role in RNA export and in tethering Nup98 and Nup153 to the nucleus. NUP160 is involved in poly(A)+ RNA transport.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NUP160
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR