Tisk…

Anti-TTK Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-7850R-CY7EA 24190 CZK
BOSSBS-7850R-CY7
Anti-TTK Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Phosphorylates proteins on serine, threonine, and tyrosine. Probably associated with cell proliferation. Essential for chromosome alignment by enhancing AURKB activity (via direct CDCA8 phosphorylation) at the centromere, and for the mitotic checkpoint.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TTK
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR