Tisk…

Anti-ZWINT Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BOSSBS-7852R-CY5EA 24190 CZK
BOSSBS-7852R-CY5
Anti-ZWINT Rabbit Polyclonal Antibody (Cy5®)
Protilátky
ZWINT is a component of the MIS12 complex, which is required for kinetochore function and spindle checkpoint activity. ZWINT interacts with ZW10 and targets it to the kinetochore at prometaphase. ZWINT is detectable on the kinetochore till late anaphase. Inhibition of ZWINT expression results in chromosome missegregation, spindle checkpoint failure and, ultimately, cell death upon cytokinesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZWINT
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR