Tisk…

Anti-CCNYL1 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BOSSBS-7889R-CY3EA 24190 CZK
BOSSBS-7889R-CY3
Anti-CCNYL1 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
CCNYL1 belongs to the cyclin family, Cyclin Y subfamily and contains 1 cyclin N terminal domain. The specific function of CCNYL1 is not yet known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCNYL1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR