Tisk…

Anti-DUSP26 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-7910R-A647EA 24190 CZK
BOSSBS-7910R-A647
Anti-DUSP26 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Inactivates MAPK1 and MAPK3 which leads to dephosphorylation of heat shock factor protein 4 and a reduction in its DNA-binding activity. Inhibits MAP kinase p38 by dephosphorylating it and inhibits p38-mediated apoptosis in anaplastic thyroid cancer cells. Can also induce activation of MAP kinase p38 and c-Jun N-terminal kinase (JNK).Tissue specificity:Brain. In the brain it is expressed ubiquitously except in the hippocampus. Expressed in embryonal cancers (retinoblastoma, neuroepithilioma and neuroblastoma) and in anaplatic thyroid cancer.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DUSP26
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR