Tisk…

Anti-GAK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BOSSBS-7912R-FITCEA 24190 CZK
BOSSBS-7912R-FITC
Anti-GAK Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Associates with cyclin G and CDK5. Seems to act as an auxilin homolog that is involved in the uncoating of clathrin-coated vesicles by Hsc70 in non-neuronal cells. Expression oscillates slightly during the cell cycle, peaking at G1.Tissue specificity: Ubiquitous. Highest in testis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GAK protein
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR