Tisk…

Anti-SRPK2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-7923R-A647EA 24190 CZK
BOSSBS-7923R-A647
Anti-SRPK2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
SRPK2 belongs to the protein kinase superfamily. It phosphorylates RS domain-containing proteins, such as SFRS1 and SFRS2 on serine residues. It has a role in spliceosome assembly and in mediating the trafficking of splicing factors and appears to mediate HBV core protein phosphorylation which is a prerequisite for pregenomic RNA encapsidation into viral capsids. SRPK2 highly expressed in brain, moderately expressed in heart and skeletal muscle and at low levels in lung, liver, and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRPK2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR