Tisk…

Anti-SIPA1L2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-7927R-HRPEA 24190 CZK
BOSSBS-7927R-HRP
Anti-SIPA1L2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
SIPA1L2

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SIPA1L2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR