Tisk…

Anti-AOC1 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BOSSBS-7949R-CY5.5EA 24190 CZK
BOSSBS-7949R-CY5.5
Anti-AOC1 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Catalyzes the degradation of compounds such as putrescine, histamine, spermine, and spermidine, substances involved in allergic and immune responses, cell proliferation, tissue differentiation, tumor formation, and possibly apoptosis. Placental DAO is thought to play a role in the regulation of the female reproductive function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AOC1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR