Tisk…

Anti-AMN1 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-7960R-CY7EA 24190 CZK
BOSSBS-7960R-CY7
Anti-AMN1 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
AMN1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AMN1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR