Tisk…

Anti-CMTM5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BOSSBS-8023R-A555EA 24190 CZK
BOSSBS-8023R-A555
Anti-CMTM5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
This gene belongs to the chemokine-like factor gene superfamily. This family of genes encodes multi-pass membrane proteins that are similar to both the chemokine and the transmembrane 4 superfamilies of signaling molecules. Alternate transcriptional splice variants of this gene, encoding different isoforms, have been characterized. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CMTM5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR