Tisk…

Anti-CMTM7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-8026R-A647EA 24190 CZK
BOSSBS-8026R-A647
Anti-CMTM7 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. The protein encoded by this gene is highly expressed in leukocytes, but its exact function is unknown. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CMTM7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR