Tisk…

Anti-CNKSR1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-8028R-HRPEA 24190 CZK
BOSSBS-8028R-HRP
Anti-CNKSR1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
This gene is a necessary element in receptor tyrosine kinase pathways, possibly as a tyrosine phosphorylation target. It is involved in regulation of RAF in the MAPK pathway and may also play a role in a MAPK-independent pathway. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CNKSR1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR