Tisk…

Anti-COLEC10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BOSSBS-8033R-A488EA 24190 CZK
BOSSBS-8033R-A488
Anti-COLEC10 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
Lectin that binds to various sugars: galactose >mannose = fucose >N-acetylglucosamine >N-acetylgalactosamine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COLEC10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR