Tisk…

Anti-COMMD1 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BOSSBS-8034R-CY3EA 24190 CZK
BOSSBS-8034R-CY3
Anti-COMMD1 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
Promotes ubiquitination of NF-kappa-B subunit RELA and its subsequent proteasomal degradation. Down-regulates NF-kappa-B activity. Down-regulates SOD1 activity by interfering with its homodimerization. Plays a role in copper ion homeostasis. Can bind one copper ion per monomer. May function to facilitate biliary copper excretion within hepatocytes.Tissue specificity:Ubiquitous. Highest expression in the liver, with lower expression in brain, lung, placenta, pancreas, small intestine, heart, skeletal muscle, kidney and placenta.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COMMD1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR