Tisk…

Anti-KLHL14 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BOSSBS-8056R-FITCEA 24190 CZK
BOSSBS-8056R-FITC
Anti-KLHL14 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
KLHL14 is a member of the KLHL family. The kelch motif was initially discovered in Kelch. In this protein there are six copies of the motif. It has been shown for one member that it is related to Galactose Oxidase for which a structure has been solved. Two isoforms of this protein exist - 70kDa and 43kDa.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLHL14
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR