Tisk…

Anti-SRRM1 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-8066R-CY7EA 24190 CZK
BOSSBS-8066R-CY7
Anti-SRRM1 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Part of pre- and post-splicing multiprotein mRNP complexes. Involved in numerous pre-mRNA processing events. Promotes constitutive and exonic splicing enhancer (ESE)-dependent splicing activation by bridging together sequence-specific (SR family proteins, SFRS4, SFRS5 and TRA2B/SFRS10) and basal snRNP (SNRP70 and SNRPA1) factors of the spliceosome. Stimulates mRNA 3'-end cleavage independently of the formation of an exon junction complex. Binds both pre-mRNA and spliced mRNA 20-25 nt upstream of exon-exon junctions. Binds RNA and DNA with low sequence specificity and has similar preference for either double- or single-stranded nucleic acid substrates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SRRM1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR