Tisk…

Anti-CCDC124 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-8083R-A647EA 24190 CZK
BOSSBS-8083R-A647
Anti-CCDC124 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
CCDC124

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC124
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR