Tisk…

Anti-CCDC129 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BOSSBS-8107R-CY3EA 24190 CZK
BOSSBS-8107R-CY3
Anti-CCDC129 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
CCDC129

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC129
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR