Tisk…

Anti-CFAP58 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-8118R-HRPEA 24190 CZK
BOSSBS-8118R-HRP
Anti-CFAP58 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
CCDC147

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CFAP58
Clonality: Polyclonal
Clone: 0
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR