Tisk…

Anti-CCDC158 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BOSSBS-8122R-CY5.5EA 24190 CZK
BOSSBS-8122R-CY5.5
Anti-CCDC158 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
CCDC158

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC158
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR