Tisk…

Anti-CCDC25 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-8125R-A647EA 24190 CZK
BOSSBS-8125R-A647
Anti-CCDC25 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
CCDC25 is a 208 amino acid protein encoded by a gene that maps to human chromosome 8p21.1. Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certain facial and developmental defects. WRN is a DNA helicase encoded by chromosome 8 and shown defective in those with the early aging disorder Werner syndrome. Chromosome 8 is also associated with Pfeiffer syndrome, congenital hypothyroidism and Waardenburg syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC25
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR