Tisk…

Anti-CCDC28B Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-8128R-CY7EA 24190 CZK
BOSSBS-8128R-CY7
Anti-CCDC28B Rabbit Polyclonal Antibody (Cy7®)
Protilátky
This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, adevelopmental disorder caused by deletion of multiple genes at 7q11.23. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CCDC28B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR