Tisk…

Anti-CEP112 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-8133R-HRPEA 24190 CZK
BOSSBS-8133R-HRP
Anti-CEP112 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
This gene encodes a protein with filament, myosin tail and ATPase domains. Orthologs of this gene exist in mouse, rat and chimp. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq, Jul 2008].

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CEP112
Clonality: Polyclonal
Clone: 0
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR