Tisk…

Anti-GOLGA2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-8155R-A647EA 24190 CZK
BOSSBS-8155R-A647
Anti-GOLGA2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Golgi auto-antigen; probably involved in maintaining cis-Golgi structure.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GM130
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR