Tisk…

Anti-COX5A Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BOSSBS-8166R-A350EA 24190 CZK
BOSSBS-8166R-A350
Anti-COX5A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer of proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit Va of the human mitochondrial respiratory chain enzyme. A pseudogene COX5AP1 has been found in chromosome 14q22. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COX5A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR