Tisk…

Anti-GBL Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BOSSBS-8161R-A350EA 24190 CZK
BOSSBS-8161R-A350
Anti-GBL Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Subunit of both mTORC1 and mTORC2, which regulates cell growth and survival in response to nutrient and hormonal signals. mTORC1 is activated in response to growth factors or amino-acids. Growth factor-stimulated mTORC1 activation involves a AKT1-mediated phosphorylation of TSC1-TSC2, which leads to the activation of the RHEB GTPase that potently activates the protein kinase activity of mTORC1. Amino-acid-signaling to mTORC1 requires its relocalization to the lysosomes mediated by the Ragulator complex and the Rag GTPases. Activated mTORC1 up-regulates protein synthesis by phosphorylating key regulators of mRNA translation and ribosome synthesis. mTORC1 phosphorylates EIF4EBP1 and releases it from inhibiting the elongation initiation factor 4E (eiF4E). mTORC1 phosphorylates and activates S6K1 at 'Thr-389', which then promotes protein synthesis by phosphorylating PDCD4 and targeting it for degradation.Tissue specificity:Broadly expressed, with highest levels in skeletal muscle, heart and kidney.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G protein beta subunit like
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR