Tisk…

Anti-CHD3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BOSSBS-8162R-FITCEA 24190 CZK
BOSSBS-8162R-FITC
Anti-CHD3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Component of the histone deacetylase NuRD complex which participates in the remodeling of chromatin by deacetylating histones. Required for anchoring centrosomal pericentrin in both interphase and mitosis, for spindle organization and centrosome integrity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHD3
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR