Tisk…

Anti-CHCHD6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BOSSBS-8167R-FITCEA 24190 CZK
BOSSBS-8167R-FITC
Anti-CHCHD6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
CHCHD6

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHCHD6
Clonality: Polyclonal
Clone: 0
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR