Tisk…

Anti-KIF11 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BOSSBS-8177R-FITCEA 24190 CZK
BOSSBS-8177R-FITC
Anti-KIF11 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
Motor protein required for establishing a bipolar spindle. Blocking of KIF11 prevents centrosome migration and arrest cells in mitosis with monoastral microtubule arrays.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIF11
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR