Tisk…

Anti-NEEP21 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-8204R-CY7EA 24190 CZK
BOSSBS-8204R-CY7
Anti-NEEP21 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
NEEP21, also known as Brain neuron cytoplasmic protein 1, NSG1 (neuron-specific proteins family member 1), P21 or D4S234E, is a single pass type II membrane protein belonging to the NSG family. It is highly expressed during neuronal maturation but its expression is downregulated in adult tissues. NEEP21 predominantly localizes to Rab 4-positive early endosomes in the somatodendritic neuronal compartment and is essential for proper receptor sorting and recycling in neurons. It associates with GRIP1 and GluR-2 and mediates the surface expression of GluR-2. When this interaction is interrupted, GluR-2 accumulates in early endosomes and leads to changes in evoked synaptic current properties. In addition, NEEP21 forms a complex with the SNARE protein, Syntaxin 13 (also known as Syntaxin 12), and participates in the recycling of transferrin receptors (TFRs) and NTR2 (neurotensin receptor 2).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NEEP21
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR