Tisk…

Anti-FAM50A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Dodavatel: Bioss
BOSSBS-8208R-A555EA 24190 CZK
BOSSBS-8208R-A555
Anti-FAM50A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Protilátky
FAM50A (Family with sequence similarity 50, member A) is a basic protein containing a nuclear localization signal. Its specifc function is unknown. It may be a DNA-binding protein or transcriptional factor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM50A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR