Tisk…

Anti-FAM155A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BOSSBS-8223R-A488EA 24190 CZK
BOSSBS-8223R-A488
Anti-FAM155A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
FAM155A

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FAM155A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR