Tisk…

Anti-FRMD1 Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BOSSBS-8232R-CY5EA 24190 CZK
BOSSBS-8232R-CY5
Anti-FRMD1 Rabbit Polyclonal Antibody (Cy5®)
Protilátky
No data available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FRMD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR