Tisk…

Anti-GIMAP4 Rabbit Polyclonal Antibody (Cy7®)

Dodavatel: Bioss
BOSSBS-8270R-CY7EA 24190 CZK
BOSSBS-8270R-CY7
Anti-GIMAP4 Rabbit Polyclonal Antibody (Cy7®)
Protilátky
Exhibits intrisinic GTPase activity. Shows a higher affinity for GDP over GTP (about 12-fold higher), and binding shows an absolute requirement for magnesium.Tissue specificity:Highly expressed in spleen and peripheral blood leukocytes that contain mostly T- and B-lymphocytes. Expressed specifically in resting T- and B-lymphocytes and expression significantly decreases during B- or T-lymphocyte activation. Expressed at lower levels in thymus, ovary, colon and small intestine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIMAP4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR