Tisk…

Anti-GIMAP6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-8272R-A647EA 24190 CZK
BOSSBS-8272R-A647
Anti-GIMAP6 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Highly expressed in spleen, lymph nodes, lung and placenta. Expressed at moderate level in thymus, kidney, heart and digestive tract. Weakly expressed in other lymphoid tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIMAP6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR