Tisk…

Anti-GIMAP7 Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BOSSBS-8273R-CY3EA 24190 CZK
BOSSBS-8273R-CY3
Anti-GIMAP7 Rabbit Polyclonal Antibody (Cy3®)
Protilátky
The GTPase of the immunity-associated protein (GIMAP) family of proteins include seven members that are expressed by genes residing on human chromosome 7. GIMAP proteins have been implicated in the regulation of lymphomyeloid cell survival. GIMAP7 (GTPase IMAP family member 7), also known as IAN7 (immunity-associated nucleotide 7), is a 300 amino acid protein encoded by a gene that maps to human chromosome 7q36.1. Chromosome 7 has been linked to Osteogenesis imperfecta, Pendred syndrome, Lissencephaly, Citrullinemia and Shwachman-Diamond syndrome. The deletion of a portion of the q arm of chromosome 7 is associated with Williams-Beuren syndrome, a condition characterized by mild mental retardation, an unusual comfort and friendliness with strangers and an elfin appearance. Deletions of portions of the q arm of chromosome 7 are also seen in a number of myeloid disorders including cases of acute myelogenous leukemia and myelodysplasia.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GIMAP7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR