Tisk…

Anti-TXNRD1 Rabbit Polyclonal Antibody (Cy5.5®)

Dodavatel: Bioss
BOSSBS-8299R-CY5.5EA 24780 CZK
BOSSBS-8299R-CY5.5
Anti-TXNRD1 Rabbit Polyclonal Antibody (Cy5.5®)
Protilátky
Isoform 1 may possess glutaredoxin activity as well as thioredoxin reductase activity and induces actin and tubulin polymerization, leading to formation of cell membrane protrusions. Isoform 4 enhances the transcriptional activity of estrogen receptors alpha and beta while isoform 5 enhances the transcriptional activity of the beta receptor only. Isoform 5 also mediates cell death induced by a combination of interferon-beta and retinoic acid.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TXNRD1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR