Tisk…

Anti-PRDX2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BOSSBS-8300R-A350EA 24190 CZK
BOSSBS-8300R-A350
Anti-PRDX2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Peroxiredoxin (Prx) is an antioxidant enzyme detoxifying reactive oxygen species and has a cysteine at the active site. Prx enzymes modulate various receptor signaling pathways and protect cells from oxidatively induced death. Peroxiredoxin 1 to 4 have two conserved Cys residues corresponding to Cys51 and Cys172 of mammalian Peroxiredoxin 1. The active site cysteine(Cys51) is oxidized to cysteine sulfenic acid(Cys51-SOH) when a peroxide is reduced. Because Cys51-SOH is unstable, it forms a disulfide with Cys172-SH which comes from the other subunit of the homodimer. The disulfide is then reduced back to the Prx active thiol form by the thioredoxin-thioredoxin reductase system. However, the formation of the disulfide is a slow process. Thus under oxidative stress conditions, the sulfenic intermediate(Cys51-SOH) can be easily over oxidized to cysteine sulfinic acid(Cys-SO2H) or cysteine sulfonic acid(Cys-SO3H) before it is able to form a disulfide. Recent studies suggest that over oxidized Prx can be reduced back to the active form during recovery after oxidative stress.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRDX2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR