Tisk…

Anti-CFP Rabbit Polyclonal Antibody (Cy3®)

Dodavatel: Bioss
BOSSBS-8306R-CY3EA 24100 CZK
BOSSBS-8306R-CY3
Anti-CFP Rabbit Polyclonal Antibody (Cy3®)
Protilátky
A positive regulator of the alternate pathway of complement. It binds to and stabilizes the C3- and C5-convertase enzyme complexes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CFP
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR