Tisk…

Anti-PANK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Dodavatel: Bioss
BOSSBS-8340R-A488EA 24190 CZK
BOSSBS-8340R-A488
Anti-PANK4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Protilátky
PANK4 belongs to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the physiological regulation of the intracellular coenzyme A (CoA) concentration. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PANK4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR