Tisk…

Anti-UBE2Q1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-8383R-HRPEA 24190 CZK
BOSSBS-8383R-HRP
Anti-UBE2Q1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (UBE1s), ubiquitin-conjugating enzymes (UBE2s), and ubiquitin-protein ligases (UBE3s). When ubiquitin is activated by a UBE1, it is transferred to the cysteine residue on a UBE2. UBE2 then binds a UBE3, which transfers the ubiquitin from the UBE2 cysteine to a lysine residue on the target protein. Ubiquitin-conjugating enzyme E2 Q1 (UBE2Q1), also known as ubiquitin-protein ligase Q1 or ubiquitin carrier protein Q1, is an 422 amino acid protein belonging to the ubiquitin-conjugating enzyme (UBE2) family. Two named isoforms of UBE2Q1 exist as a result of alternative splicing events.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: UBE2Q1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR