Tisk…

Anti-LIN28A/B Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BOSSBS-8443R-CY5EA 24190 CZK
BOSSBS-8443R-CY5
Anti-LIN28A/B Rabbit Polyclonal Antibody (Cy5®)
Protilátky
LIN-28 is a highly conserved, RNA-binding, cytoplasmic protein. It consists of a cold shock domain and retroviral-type (CCHC) zinc finger motifs that were first identified in Caenorhabditis elegans. LIN-28 controls the timing of events during embryonic development and is readily expressed in embryos, embryonic stem cells and embryonal carcinoma cells. The presence of LIN-28 persists in some adult tissues including cardiac and skeletal muscle. In differentiating myoblasts, LIN-28 increases protein synthesis efficiency and binds to the growth and differentiation factor IGF-II.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LIN28A/B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR