Tisk…

Anti-SNAPC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dodavatel: Bioss
BOSSBS-8452R-FITCEA 24190 CZK
BOSSBS-8452R-FITC
Anti-SNAPC3 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Protilátky
SNAPC3 is part of the SNAPc complex required for the transcription of both RNA polymerase II and III small-nuclear RNA genes. SNAPC3 binds to the proximal sequence element (PSE), a non-TATA-box basal promoter element common to these 2 types of genes. SNAPC3 recruits TBP and BRF2 to the U6 snRNA TATA box.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNAPC3
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR