Tisk…

Anti-SENP5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dodavatel: Bioss
BOSSBS-8464R-HRPEA 24190 CZK
BOSSBS-8464R-HRP
Anti-SENP5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Protilátky
Protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO3 to its mature form and deconjugation of SUMO2 and SUMO3 from targeted proteins. Has weak proteolytic activity against full-length SUMO1 or SUMO1 conjugates. Required for cell division.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SENP5
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR