Tisk…

Anti-FBXL16 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Dodavatel: Bioss
BOSSBS-8474R-A647EA 24190 CZK
BOSSBS-8474R-A647
Anti-FBXL16 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Protilátky
Substrate-recognition component of the SCF (SKP1-CUL1-F-box protein)-type E3 ubiquitin ligase complex.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXL16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR