Tisk…

Anti-FBXO33 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Dodavatel: Bioss
BOSSBS-8471R-A350EA 24190 CZK
BOSSBS-8471R-A350
Anti-FBXO33 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Protilátky
Members of the F-box protein family, such as FBXO33, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1, cullin and F-box proteins act as protein-ubiquitin ligases and mediates the ubiquitination and subsequent proteasomal degradation of target proteins. FBXO33 probably recognizes and binds to phosphorylated target proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FBXO33
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR