Tisk…

Anti-RFPL4B Rabbit Polyclonal Antibody (Cy5®)

Dodavatel: Bioss
BOSSBS-8495R-CY5EA 24190 CZK
BOSSBS-8495R-CY5
Anti-RFPL4B Rabbit Polyclonal Antibody (Cy5®)
Protilátky
RFPL4B contains 1 B30.2/SPRY domain and 1 RING type zinc finger. The function of RFPL4B remains unknown.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RFPL4B
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR